5-Brom-2-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-4-bromobenzotrifluoride

AS31345G1 AS31345G5
ACTIAS31345G1EA 0 CHF
ACTIAS31345G1 ACTIAS31345G5
5-Brom-2-(trifluormethyl)anilin
5-Brom-2-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MW: 240.0200 g/mol
MDL Number: MFCD00082853
CAS-Nummer: 703-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR