3-Brom-5-fluor-2-methylanilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-6-bromo-4-fluorotoluene

AS21879G1 AS21879G5
ACTIAS21879G1EA 21 CHF
ACTIAS21879G1 ACTIAS21879G5
3-Brom-5-fluor-2-methylanilin
3-Brom-5-fluor-2-methylanilin
Formel: C₇H₇BrFN
MW: 204.0400 g/mol
MDL Number: MFCD03094193
CAS-Nummer: 502496-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR