3-Brom-4-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS36173G1 AS36173G5
ACTIAS36173G1EA 0 CHF
ACTIAS36173G1 ACTIAS36173G5
3-Brom-4-(trifluormethyl)anilin
3-Brom-4-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MW: 240.0200 g/mol
MDL Number: MFCD12828280
CAS-Nummer: 172215-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR