1-Brom-3-(difluormethyl)benzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-3-(difluoromethyl)benzene, 3-Bromobenzal fluoride

AS34465G1 AS34465G5
ACTIAS34465G1EA 0 CHF
ACTIAS34465G1 ACTIAS34465G5
1-Brom-3-(difluormethyl)benzol
1-Bromo-3-(difluoromethyl)benzene
Formel: C₇H₅BrF₂
MDL Number: MFCD07368984
CAS-Nummer: 29848-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR