3-Chlor-4-isopropoxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35196G1 AS35196G10 AS35196G5
ACTIAS35196G1EA 0 CHF
ACTIAS35196G1 ACTIAS35196G10 ACTIAS35196G5
3-Chlor-4-isopropoxyphenylboronsäure
3-Chlor-4-isopropoxyphenylboronsäure
Formel: C₉H₁₂BClO₃
MW: 214,4500 g/mol
MDL Number: MFCD04974065
CAS-Nummer: 480438-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR