2,4-Dimethoxyphenol

Lieferant: ACTIVATE SCIENTIFIC

AS33696G1 AS33696G5
ACTIAS33696G1EA 0 CHF
ACTIAS33696G1 ACTIAS33696G5
2,4-Dimethoxyphenol
2,4-Dimethoxyphenol
Formel: C₈H₁₀O₃
MW: 154,1632 g/mol
MDL Number: MFCD00060637
CAS-Nummer: 13330-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR