2-Amino-4-(trifluormethyl)benzonitril
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Amino-4-cyanobenzotrifluoride
AS35156G1
AS35156G5
ACTIAS35156G1EA
0
CHF
ACTIAS35156G1
ACTIAS35156G5
2-Amino-4-(trifluormethyl)benzonitril
2-Amino-4-(trifluormethyl)benzonitril
Formel:
C₈H₅F₃N₂ MW: 186,1339 g/mol |
MDL Number:
MFCD04974124 CAS-Nummer: 1483-54-1 EINECS: 216-047-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...