4-(Brommethyl)-3-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35088G1 AS35088G5
ACTIAS35088G1EA 0 CHF
ACTIAS35088G1 ACTIAS35088G5
4-(Brommethyl)-3-fluorbenzonitril
4-(Brommethyl)-3-fluorbenzonitril
Formel: C₈H₅BrFN
MDL Number: MFCD07368341
CAS-Nummer: 105942-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR