4-Chlor-3-methoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS35127G1 AS35127G5
ACTIAS35127G1EA 0 CHF
ACTIAS35127G1 ACTIAS35127G5
4-Chlor-3-methoxybenzaldehyd
4-Chloro-3-methoxybenzaldehyde
Formel: C₈H₇ClO₂
MW: 170,5900 g/mol
MDL Number: MFCD07787490
CAS-Nummer: 13726-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR