3-Chlor-2-methoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30791G1 AS30791G5
ACTIAS30791G1EA 0 CHF
ACTIAS30791G1 ACTIAS30791G5
3-Chlor-2-methoxybenzaldehyd
3-Chloro-2-methoxybenzaldehyde
Formel: C₈H₇ClO₂
MW: 170,5930 g/mol
MDL Number: MFCD09999135
CAS-Nummer: 223778-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR