2-Chlor-6-methoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31657G1 AS31657G5
ACTIAS31657G1EA 0 CHF
ACTIAS31657G1 ACTIAS31657G5
2-Chlor-6-methoxybenzaldehyd
2-Chloro-6-methoxybenzaldehyde
Formel: C₈H₇ClO₂
MW: 170,5900 g/mol
MDL Number: MFCD08059233
CAS-Nummer: 29866-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR