3-Brom-5-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS32459G1 AS32459G5
ACTIAS32459G1EA 0 CHF
ACTIAS32459G1 ACTIAS32459G5
3-Brom-5-fluoranilin
3-Brom-5-fluoranilin
Formel: C₆H₅BrFN
MW: 190,01 g/mol
MDL Number: MFCD07779529
CAS-Nummer: 134168-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR