5-Brom-2,4-difluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS35887G1 AS35887G5
ACTIAS35887G1EA 0 CHF
ACTIAS35887G1 ACTIAS35887G5
5-Brom-2,4-difluoranilin
5-Brom-2,4-difluoranilin
Formel: C₆H₄BrF₂N
MW: 208,0000 g/mol
MDL Number: MFCD03426368
CAS-Nummer: 452-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR