2-Brom-4,5-difluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32445G1 AS32445G5
ACTIAS32445G1EA 0 CHF
ACTIAS32445G1 ACTIAS32445G5
2-Brom-4,5-difluorbenzonitril
2-Brom-4,5-difluorbenzonitril
Formel: C₇H₂BrF₂N
MDL Number: MFCD09864661
CAS-Nummer: 64695-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR