3-Bromo-2-chlorophenol
Lieferant: ACTIVATE SCIENTIFIC
AS33337G1
AS33337G5
ACTIAS33337G1EA
0
CHF
ACTIAS33337G1
ACTIAS33337G5
3-Bromo-2-chlorophenol
3-Bromo-2-chlorophenol
Formel:
C₆H₄BrClO MW: 207,452 g/mol |
MDL Number:
MFCD08166321 CAS-Nummer: 863870-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...