5-Cyan-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33423G1 AS33423G5
ACTIAS33423G1EA 0 CHF
ACTIAS33423G1 ACTIAS33423G5
5-Cyan-2-methylpyridin
5-Cyan-2-methylpyridin
Formel: C₇H₆N₂
MDL Number: MFCD00038042
CAS-Nummer: 3222-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR