2-Fluor-3-nitrobenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2-Fluoro-3-nitrophenyl)methanol

AS31525G1 AS31525G5
ACTIAS31525G1EA 0 CHF
ACTIAS31525G1 ACTIAS31525G5
2-Fluor-3-nitrobenzylalkohol
2-Fluor-3-nitrobenzylalkohol
Formel: C₇H₆FNO₃
MDL Number: MFCD09835208
CAS-Nummer: 946126-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR