4-Chlor-3-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS22607G1 AS22607G5
ACTIAS22607G1EA 0 CHF
ACTIAS22607G1 ACTIAS22607G5
4-Chlor-3-methoxybenzonitril
4-Chlor-3-methoxybenzonitril
Formel: C₈H₆ClNO
MDL Number: MFCD09753729
CAS-Nummer: 189628-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR