2-Chlor-6-methoxybenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS30831G1 AS30831G5
ACTIAS30831G1EA 0 CHF
ACTIAS30831G1 ACTIAS30831G5
2-Chlor-6-methoxybenzonitril
2-Chlor-6-methoxybenzonitril
Formel: C₈H₆ClNO
MDL Number: MFCD00019743
CAS-Nummer: 6575-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR