5-Brom-2-chlor-4-fluorphenol

Lieferant: ACTIVATE SCIENTIFIC

AS32262G1 AS32262G5
ACTIAS32262G1EA 0 CHF
ACTIAS32262G1 ACTIAS32262G5
5-Brom-2-chlor-4-fluorphenol
5-Brom-2-chlor-4-fluorphenol
Formel: C₆H₃BrClFO
MW: 225,4428 g/mol
MDL Number: MFCD08436979
CAS-Nummer: 148254-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR