5-Brombenzimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS23481G1 AS23481G5
ACTIAS23481G1EA 0 CHF
ACTIAS23481G1 ACTIAS23481G5
5-Brombenzimidazol
5-Bromobenzimidazole
Formel: C₇H₅BrN₂
MDL Number: MFCD00160001
CAS-Nummer: 4887-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR