2-Chlor-4-fluor-3-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32063G1 AS32063G5
ACTIAS32063G1EA 0 CHF
ACTIAS32063G1 ACTIAS32063G5
2-Chlor-4-fluor-3-methylbenzonitril
2-Chlor-4-fluor-3-methylbenzonitril
Formel: C₈H₅ClFN
MW: 169,5834 g/mol
MDL Number: MFCD13193544
CAS-Nummer: 796600-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR