2-Acetylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Acetylbenzenecarbonitrile

AS34434G1 AS34434G5
ACTIAS34434G1EA 0 CHF
ACTIAS34434G1 ACTIAS34434G5
2-Acetylbenzonitril
2-Acetylbenzonitril
Formel: C₉H₇NO
MDL Number: MFCD00272571
CAS-Nummer: 91054-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR