3-Nitro-4-chinolinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Hydroxy-3-nitroquinoline, 3-Nitroquinolin-4-ol

AS35856G1 AS35856G5
ACTIAS35856G1EA 0 CHF
ACTIAS35856G1 ACTIAS35856G5
3-Nitro-4-chinolinol
3-Nitro-4-chinolinol
Formel: C₉H₆N₂O₃
MW: 190,1600 g/mol
MDL Number: MFCD00277722
CAS-Nummer: 50332-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR