3-Fluor-2-nitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32653G1EA 0 CHF
ACTIAS32653G1 ACTIAS32653G5
3-Fluor-2-nitrobenzonitril
3-Fluor-2-nitrobenzonitril
Formel: C₇H₃FN₂O₂
MW: 166,1100 g/mol
MDL Number: MFCD09864664
CAS-Nummer: 1000339-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR