3-Cyan-4-methylanilin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Amino-2-methylbenzonitrile
AS21392G100
AS21392G25
ACTIAS21392G100EA
0
CHF
ACTIAS21392G100
ACTIAS21392G25
3-Cyan-4-methylanilin
3-Cyano-4-methylaniline
Formel:
C₈H₈N₂ MW: 132,16 g/mol |
MDL Number:
MFCD00017608 CAS-Nummer: 50670-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...