5-Nitro-m-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methyl-5-nitrobenzonitrile

AS36299G1 AS36299G5
ACTIAS36299G1EA 0 CHF
ACTIAS36299G1 ACTIAS36299G5
5-Nitro-m-toluonitril
5-Nitro-m-toluonitril
Formel: C₈H₆N₂O₂
MW: 162,150 g/mol
MDL Number: MFCD17170423
CAS-Nummer: 124289-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR