3-Methoxypicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Methoxypyridine-2-carboxylic acid

AS34593G1 AS34593G5
ACTIAS34593G1EA 0 CHF
ACTIAS34593G1 ACTIAS34593G5
3-Methoxypicolinsäure
3-Methoxypicolinsäure
Formel: C₇H₇NO₃
MW: 153,1354 g/mol
MDL Number: MFCD07367569
CAS-Nummer: 16478-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR