6-Methylpicolinsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid
AS21229G25
AS21229G5
ACTIAS21229G25EA
95
CHF
ACTIAS21229G25
ACTIAS21229G5
6-Methylpicolinsäure
6-Methylpicolinsäure
Formel:
C₇H₇NO₂ MW: 137.14 g/mol |
MDL Number:
MFCD00023481 CAS-Nummer: 934-60-1 EINECS: 213-287-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...