2-Methyl-1-(tributylstannyl)-1-propen

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51291G1 AS51291G5
ACTIAS51291G1EA 156 CHF
ACTIAS51291G1 ACTIAS51291G5
2-Methyl-1-(tributylstannyl)-1-propen
2-Methyl-1-(tributylstannyl)-1-propen
Formel: C₁₆H₃₄Sn
MDL Number: MFCD09750019
CAS-Nummer: 66680-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR