3-(3-Methoxyphenoxy)propionsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(3-Methoxyphenoxy)propionic Acid

AS22925G1 AS22925G5
ACTIAS22925G1EA 108 CHF
ACTIAS22925G1 ACTIAS22925G5
3-(3-Methoxyphenoxy)propionsäure
3-(3-Methoxyphenoxy)propionsäure
Formel: C₁₀H₁₂O₄
MW: 196,1999 g/mol
MDL Number: MFCD00045943
CAS-Nummer: 49855-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR