2-(3-Methoxyphenoxy)propionsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(3-Methoxyphenoxy)propionic Acid
AS23600G1
AS23600G25
AS23600G5
ACTIAS23600G1EA
115
CHF
ACTIAS23600G1
ACTIAS23600G25
ACTIAS23600G5
2-(3-Methoxyphenoxy)propionsäure
2-(3-Methoxyphenoxy)propionsäure
Formel:
C₁₀H₁₂O₄ MW: 196.1999 g/mol |
MDL Number:
MFCD01931912 CAS-Nummer: 7309-52-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...