2-Methoxy-6-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32647G1 AS32647G5
ACTIAS32647G1EA 0 CHF
ACTIAS32647G1 ACTIAS32647G5
2-Methoxy-6-methylbenzonitril
2-Methoxy-6-methylbenzonitril
Formel: C₉H₉NO
MW: 147,1739 g/mol
MDL Number: MFCD10699539
CAS-Nummer: 53005-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR