6-Iod-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Amino-4-iodotoluene, 2-Iodo-5-methylaniline

AS35371G1 AS35371G10 AS35371G5
ACTIAS35371G1EA 0 CHF
ACTIAS35371G1 ACTIAS35371G10 ACTIAS35371G5
6-Iod-m-toluidin
6-Iod-m-toluidin
Formel: C₇H₈IN
MW: 233.0496 g/mol
MDL Number: MFCD00833395
CAS-Nummer: 13194-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR