5-Hydroxypicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-hydroxypyridine-2-carboxylic acid

AS30959G1 AS30959G5
ACTIAS30959G1EA 0 CHF
ACTIAS30959G1 ACTIAS30959G5
5-Hydroxypicolinsäure
5-Hydroxypicolinsäure
Formel: C₆H₅NO₃
MW: 139,11 g/mol
MDL Number: MFCD00661309
CAS-Nummer: 15069-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR