5-Brom-m-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-methylbenzonitrile

AS32245G1 AS32245G5
ACTIAS32245G1EA 0 CHF
ACTIAS32245G1 ACTIAS32245G5
5-Brom-m-toluonitril
5-Brom-m-toluonitril
Formel: C₈H₆BrN
MW: 196.0400 g/mol
MDL Number: MFCD08061970
CAS-Nummer: 124289-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR