6-Brom-o-toluonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-6-methylbenzonitrile

AS34038G1 AS34038G5
ACTIAS34038G1EA 0 CHF
ACTIAS34038G1 ACTIAS34038G5
6-Brom-o-toluonitril
6-Brom-o-toluonitril
Formel: C₈H₆BrN
MW: 196.0439 g/mol
MDL Number: MFCD09834775
CAS-Nummer: 77532-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR