1,4-Diiod-2,5-dimethoxybenzol

Lieferant: ACTIVATE SCIENTIFIC

AS35379G1 AS35379G10 AS35379G5
ACTIAS35379G1EA 0 CHF
ACTIAS35379G1 ACTIAS35379G10 ACTIAS35379G5
1,4-Diiod-2,5-dimethoxybenzol
1,4-Diiodo-2,5-dimethoxybenzene
Formel: C₈H₈I₂O₂
MW: 389,9569 g/mol
MDL Number: MFCD00461369
CAS-Nummer: 51560-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR