1,5-Dimethyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC

AS51386G1 AS51386G5
ACTIAS51386G1EA 0 CHF
ACTIAS51386G1 ACTIAS51386G5
1,5-Dimethyl-1H-pyrazol
1,5-Dimethyl-1H-pyrazole
Formel: C₅H₈N₂
MW: 96,1300 g/mol
MDL Number: MFCD01646129
CAS-Nummer: 694-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR