L(+)-Isoleucinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: L-Isoleucinol, (S)-(+)-Isoleucinol, (2S,3S)-2-Amino-3-methyl-1-pentanol, (2S,3S)-2-Amino-3-methylpentan-1-ol

AS33072G1 AS33072G5
ACTIAS33072G1EA 0 CHF
ACTIAS33072G1 ACTIAS33072G5
L(+)-Isoleucinol
L(+)-Isoleucinol
Formel: C₆H₁₅NO
MW: 117,19 g/mol
MDL Number: MFCD00004731
CAS-Nummer: 24629-25-2
EINECS: 246-371-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR