4-Amino-2,6-dimethylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dimethylpyridin-4-amine

AS1367G1 AS1367G5
ACTIAS1367G1EA 41 CHF
ACTIAS1367G1 ACTIAS1367G5
4-Amino-2,6-dimethylpyridin
4-Amino-2,6-dimethylpyridine
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
MDL Number: MFCD00130078
CAS-Nummer: 3512-80-9
EINECS: 222-515-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR