2-Amino-4,6-dimethylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33842G10 AS33842G5
ACTIAS33842G10EA 0 CHF
ACTIAS33842G10 ACTIAS33842G5
2-Amino-4,6-dimethylpyridin
2-Amino-4,6-dimethylpyridine
Formel: C₇H₁₀N₂
MDL Number: MFCD00006322
CAS-Nummer: 5407-87-4
EINECS: 226-470-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR