4-Iod-2-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS32521G1 AS32521G5
ACTIAS32521G1EA 0 CHF
ACTIAS32521G1 ACTIAS32521G5
4-Iod-2-methoxypyridin
4-Iodo-2-methoxypyridine
Formel: C₆H₆INO
MDL Number: MFCD08277272
CAS-Nummer: 98197-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR