2-Iod-5-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS51812G1 AS51812G5
ACTIAS51812G1EA 0 CHF
ACTIAS51812G1 ACTIAS51812G5
2-Iod-5-methoxypyridin
2-Iodo-5-methoxypyridine
Formel: C₆H₆INO
MDL Number: MFCD02259696
CAS-Nummer: 163129-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR