3-Amino-6-fluor-2-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Fluoro-2-methylpyridin-3-amine

AS7184G1 AS7184G5
ACTIAS7184G1EA 0 CHF
ACTIAS7184G1 ACTIAS7184G5
3-Amino-6-fluor-2-methylpyridin
3-Amino-6-fluoro-2-methylpyridine
Formel: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03094949
CAS-Nummer: 28489-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR