4-Fluorisatin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Fluoroindoline-2,3-dione

AS33782G1 AS33782G5
ACTIAS33782G1EA 0 CHF
ACTIAS33782G1 ACTIAS33782G5
4-Fluorisatin
4-Fluorisatin
Formel: C₈H₄FNO₂
MW: 165,1200 g/mol
MDL Number: MFCD01175824
CAS-Nummer: 346-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR