(R)-(+)-2-Hydroxy-3-phenylpropionsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid

AS32756G1 AS32756G10 AS32756G5
ACTIAS32756G1EA 0 CHF
ACTIAS32756G1 ACTIAS32756G10 ACTIAS32756G5
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
(R)-(+)-2-Hydroxy-3-phenylpropionsäure
Formel: C₉H₁₀O₃
MW: 166,1739 g/mol
MDL Number: MFCD00078062
CAS-Nummer: 7326-19-4
EINECS: 230-803-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR