7-Nitroisatin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Nitroindoline-2,3-dione

AS35747G1 AS35747G5
ACTIAS35747G1EA 0 CHF
ACTIAS35747G1 ACTIAS35747G5
7-Nitroisatin
7-Nitroisatin
Formel: C₈H₄N₂O₄
MW: 192.1300 g/mol
MDL Number: MFCD02066502
CAS-Nummer: 112656-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR