2-Cyan-4'-methylbiphenyl

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4'-Methyl-[1,1'-biphenyl]-2-carbonitrile

AS25354G100 AS25354G25
ACTIAS25354G100EA 148 CHF
ACTIAS25354G100 ACTIAS25354G25
2-Cyan-4'-methylbiphenyl
2-Cyan-4'-methylbiphenyl
Formel: C₁₄H₁₁N
MW: 193,26 g/mol
MDL Number: MFCD00151805
CAS-Nummer: 114772-53-1
EINECS: 422-310-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR