4,6-Dichlorpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS33664G1 AS33664G5
ACTIAS33664G1EA 0 CHF
ACTIAS33664G1 ACTIAS33664G5
4,6-Dichlorpicolinsäure
4,6-Dichlorpicolinsäure
Formel: C₆H₃Cl₂NO₂
MDL Number: MFCD09910267
CAS-Nummer: 88912-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR